Assay Details
Gene Symbol: Card19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013067580.2 | XP_012923034.1 | 5 - 6 | 723 | 81 | |
XM_021248493.1 | XP_021104152.1 | 4 - 5 | 666 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
