Assay Details
Gene Symbol: Rpp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847753.3 | XP_004847810.1 | 7 - 8 | 980 | 91 | |
XM_021260719.1 | XP_021116378.1 | 6 - 7 | 857 | 91 | ||
XM_021260720.1 | XP_021116379.1 | 7 - 8 | 847 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
