Assay Details
| Gene Symbol: S100a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871443.2 | XP_004871500.2 | 2 - 3 | 299 | 92 | |
| XM_013067942.2 | XP_012923396.1 | 3 - 4 | 1496 | 92 | ||
| XM_021249405.1 | XP_021105064.1 | 2 - 3 | 198 | 92 | ||
| XM_021249406.1 | XP_021105065.1 | 2 - 3 | 219 | 92 | ||
More Information
Set Membership: |
|

S100a5
Genomic Map