Assay Details
Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850768.3 | XP_004850825.2 | 2 - 3 | 429 | 82 | |
XM_013074378.2 | XP_012929832.1 | 2 - 3 | 493 | 82 | ||
XM_013074380.2 | XP_012929834.1 | 2 - 3 | 198 | 82 | ||
XM_021263195.1 | XP_021118854.1 | 2 - 3 | 453 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
