Assay Details
Gene Symbol: Pimreg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857264.2 | XP_004857321.1 | 3 - 4 | 992 | 71 | |
XM_013076762.2 | XP_012932216.1 | 3 - 4 | 993 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
