Assay Details
| Gene Symbol: CUNH17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857255.3 | XP_004857312.1 | 1 - 2 | 903 | 98 | |
| XM_021238192.1 | XP_021093851.1 | 1 - 2 | 903 | 98 | ||
More Information
Set Membership: |
|

CUNH17orf100
Genomic Map