Assay Details
| Gene Symbol: CUNH17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238192.1 | XP_021093851.1 | 2 - 3 | 1145 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH17orf100| Gene Symbol: CUNH17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238192.1 | XP_021093851.1 | 2 - 3 | 1145 | 92 | |
Set Membership: |
|