Assay Details
| Gene Symbol: Efemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867256.3 | XP_004867313.1 | 3 - 4 | 275 | 65 | |
| XM_021246278.1 | XP_021101937.1 | 3 - 4 | 314 | 65 | ||
| XM_021246279.1 | XP_021101938.1 | 3 - 4 | 646 | 65 | ||
More Information
Set Membership: |
|

Efemp1
Genomic Map