Assay Details
| Gene Symbol: Grpel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854389.3 | XP_004854446.1 | 2 - 3 | 473 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grpel2| Gene Symbol: Grpel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854389.3 | XP_004854446.1 | 2 - 3 | 473 | 99 | |
Set Membership: |
|