Assay Details
Gene Symbol: Rab8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873305.3 | XP_004873362.1 | 3 - 4 | 304 | 68 | |
XM_013068699.2 | XP_012924153.1 | 3 - 4 | 304 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
