Assay Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859976.3 | XP_004860033.1 | 4 - 5 | 454 | 67 | |
| XM_021240395.1 | XP_021096054.1 | 3 - 4 | 267 | 67 | ||
| XM_021240396.1 | XP_021096055.1 | 3 - 4 | 403 | 67 | ||
More Information
Set Membership: |
|

Rsu1
Genomic Map