Assay Details
| Gene Symbol: Cbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845590.3 | XP_004845647.1 | 3 - 4 | 298 | 60 | |
| XM_013072480.2 | XP_012927934.1 | 3 - 4 | 296 | 60 | ||
More Information
Set Membership: |
|

Cbx6
Genomic Map