Assay Details
Gene Symbol: Hivep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871808.3 | XP_004871865.1 | 8 - 9 | 6522 | 62 | |
XM_004871809.3 | XP_004871866.1 | 6 - 7 | 6248 | 62 | ||
XM_021249941.1 | XP_021105600.1 | 8 - 9 | 6435 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
