Assay Details
Gene Symbol: Guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021259820.1 | XP_021115479.1 | 4 - 5 | 788 | 61 | |
XM_021259821.1 | XP_021115480.1 | 4 - 5 | 630 | 61 | ||
XM_021259822.1 | XP_021115481.1 | 4 - 5 | 681 | 61 | ||
XM_021259823.1 | XP_021115482.1 | 4 - 5 | 770 | 61 | ||
XM_021259824.1 | XP_021115483.1 | 4 - 5 | 798 | 61 | ||
XM_021259825.1 | XP_021115484.1 | 4 - 5 | 668 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
