Assay Details
Gene Symbol: Bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866970.2 | XP_004867027.1 | 4 - 5 | 489 | 60 | |
XM_021245967.1 | XP_021101626.1 | 4 - 5 | 495 | 60 | ||
XM_021245968.1 | XP_021101627.1 | 4 - 5 | 478 | 60 | ||
XM_021245969.1 | XP_021101628.1 | 5 - 6 | 889 | 60 | ||
XM_021245970.1 | XP_021101629.1 | 4 - 5 | 518 | 60 | ||
XM_021245972.1 | XP_021101631.1 | 3 - 4 | 444 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
