Assay Details
Gene Symbol: LOC101715865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021240568.1 | XP_021096227.1 | 1 - 2 | 157 | 93 | |
XM_021240571.1 | XP_021096230.1 | 1 - 2 | 157 | 93 | ||
XM_021240572.1 | XP_021096231.1 | 1 - 2 | 160 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021240568.1, XM_021240571.1, XM_021240572.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
