Assay Details
Gene Symbol: Lrch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840198.2 | XP_004840255.1 | 14 - 15 | 1594 | 60 | |
XM_004840199.3 | XP_004840256.1 | 14 - 15 | 1594 | 60 | ||
XM_021254502.1 | XP_021110161.1 | 13 - 14 | 1528 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
