Assay Details
| Gene Symbol: Nr2c2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873419.3 | XP_004873476.1 | 4 - 5 | 493 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr2c2ap| Gene Symbol: Nr2c2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873419.3 | XP_004873476.1 | 4 - 5 | 493 | 111 | |
Set Membership: |
|