Assay Details
Gene Symbol: Lrrc8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004849178.3 | XP_004849235.1 | 2 - 3 | 424 | 90 | |
XM_004849179.2 | XP_004849236.1 | 2 - 3 | 170 | 90 | ||
XM_013073647.2 | XP_012929101.1 | 2 - 3 | 419 | 90 | ||
XM_013073649.2 | XP_012929103.1 | 2 - 3 | 150 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004849178.3, XM_004849179.2, XM_013073647.2, XM_013073649.2 transcripts.
Set Membership: |
![]() ![]() |
