Assay Details
| Gene Symbol: Lrrc8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849178.3 | XP_004849235.1 | 3 - 4 | 2600 | 86 | |
| XM_004849179.2 | XP_004849236.1 | 3 - 4 | 2346 | 86 | ||
| XM_013073647.2 | XP_012929101.1 | 3 - 4 | 2595 | 86 | ||
| XM_013073649.2 | XP_012929103.1 | 3 - 4 | 2326 | 86 | ||
More Information
Set Membership: |
|

Lrrc8a
Genomic Map