Assay Details
Gene Symbol: Gap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835680.3 | XP_004835737.1 | 1 - 2 | 276 | 88 | |
XM_004835681.3 | XP_004835738.1 | 1 - 2 | 179 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004835680.3, XM_004835681.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
