Assay Details
Gene Symbol: Pigg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847498.3 | XP_004847555.1 | 9 - 10 | 2213 | 96 | |
XM_013073050.2 | XP_012928504.1 | 9 - 10 | 2222 | 96 | ||
XM_013073051.2 | XP_012928505.1 | 9 - 10 | 2222 | 96 | ||
XM_013073052.2 | XP_012928506.1 | 9 - 10 | 2223 | 96 | ||
XM_013073053.2 | XP_012928507.1 | 8 - 9 | 2010 | 96 | ||
XM_013073054.2 | XP_012928508.1 | 9 - 10 | 2223 | 96 | ||
XM_021260463.1 | XP_021116122.1 | 9 - 10 | 2223 | 96 | ||
XM_021260464.1 | XP_021116123.1 | 9 - 10 | 2095 | 96 | ||
XM_021260465.1 | XP_021116124.1 | 5 - 6 | 3176 | 96 | ||
XM_021260466.1 | XP_021116125.1 | 4 - 5 | 2270 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
