Assay Details
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837973.2 | XP_004838030.1 | 15 - 16 | 1804 | 67 | |
XM_004837974.2 | XP_004838031.1 | 14 - 15 | 1792 | 67 | ||
XM_004837976.1 | XP_004838033.1 | 13 - 14 | 1675 | 67 | ||
XM_004837978.2 | XP_004838035.1 | 14 - 15 | 1645 | 67 | ||
XM_004837979.2 | XP_004838036.1 | 13 - 14 | 1633 | 67 | ||
XM_004837981.2 | XP_004838038.1 | 15 - 16 | 1656 | 67 | ||
XM_004837982.2 | XP_004838039.1 | 13 - 14 | 1558 | 67 | ||
XM_021252222.1 | XP_021107881.1 | 13 - 14 | 1672 | 67 | ||
XM_021252224.1 | XP_021107883.1 | 12 - 13 | 1663 | 67 | ||
XM_021252226.1 | XP_021107885.1 | 13 - 14 | 1636 | 67 | ||
XM_021252230.1 | XP_021107889.1 | 12 - 13 | 1555 | 67 | ||
XM_021252243.1 | XP_021107902.1 | 12 - 13 | 1480 | 67 | ||
XM_021252258.1 | XP_021107917.1 | 11 - 12 | 1360 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
