Assay Details
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021242771.1 | XP_021098430.1 | 6 - 7 | 1464 | 89 | |
XM_021242772.1 | XP_021098431.1 | 4 - 5 | 1055 | 89 | ||
XM_021242773.1 | XP_021098432.1 | 5 - 6 | 1348 | 89 | ||
XM_021242774.1 | XP_021098433.1 | 5 - 6 | 1171 | 89 | ||
XM_021242775.1 | XP_021098434.1 | 6 - 7 | 1591 | 89 | ||
XM_021242776.1 | XP_021098435.1 | 6 - 7 | 1952 | 89 | ||
XM_021242777.1 | XP_021098436.1 | 4 - 5 | 872 | 89 | ||
XM_021242778.1 | XP_021098437.1 | 3 - 4 | 821 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
