Assay Details
| Gene Symbol: Marveld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872835.2 | XP_004872892.1 | 4 - 5 | 1671 | 77 | |
| XM_021250685.1 | XP_021106344.1 | 4 - 5 | 1653 | 77 | ||
| XM_021250686.1 | XP_021106345.1 | 3 - 4 | 1635 | 77 | ||
More Information
Set Membership: |
|

Marveld2
Genomic Map