Assay Details
| Gene Symbol: LOC101702319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067620.2 | XP_012923074.1 | 32 - 33 | 4374 | 98 | |
| XM_013067621.2 | XP_012923075.1 | 32 - 33 | 4281 | 98 | ||
| XM_013067623.2 | XP_012923077.1 | 25 - 26 | 3223 | 98 | ||
More Information
Set Membership: |
|

LOC101702319
Genomic Map