Assay Details
| Gene Symbol: Gramd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854461.2 | XP_004854518.1 | 7 - 8 | 827 | 105 | |
| XM_004854463.3 | XP_004854520.2 | 5 - 6 | 2041 | 105 | ||
| XM_021266255.1 | XP_021121914.1 | 7 - 8 | 943 | 105 | ||
| XM_021266257.1 | XP_021121916.1 | 8 - 9 | 664 | 105 | ||
| XM_021266258.1 | XP_021121917.1 | 8 - 9 | 719 | 105 | ||
| XM_021266259.1 | XP_021121918.1 | 8 - 9 | 713 | 105 | ||
More Information
Set Membership: |
|

Gramd2b
Genomic Map