Assay Details
| Gene Symbol: Gramd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854461.2 | XP_004854518.1 | 8 - 9 | 915 | 79 | |
| XM_004854463.3 | XP_004854520.2 | 6 - 7 | 2129 | 79 | ||
| XM_021266255.1 | XP_021121914.1 | 8 - 9 | 1031 | 79 | ||
| XM_021266257.1 | XP_021121916.1 | 9 - 10 | 752 | 79 | ||
| XM_021266258.1 | XP_021121917.1 | 9 - 10 | 807 | 79 | ||
| XM_021266259.1 | XP_021121918.1 | 9 - 10 | 801 | 79 | ||
More Information
Set Membership: |
|

Gramd2b
Genomic Map