Assay Details
| Gene Symbol: LOC101705543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851210.3 | XP_004851267.1 | 1 - 2 | 183 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004851210.3 transcripts.
Set Membership: |
|

LOC101705543
Genomic Map