Assay Details
Gene Symbol: Cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013067570.2 | XP_012923024.1 | 5 - 6 | 786 | 74 | |
XM_021248489.1 | XP_021104148.1 | 5 - 6 | 581 | 74 | ||
XM_021248490.1 | XP_021104149.1 | 5 - 6 | 589 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
