Assay Details
Gene Symbol: LOC101719334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843641.2 | XP_004843698.1 | 1 - 2 | 338 | 56 | |
XM_013071722.2 | XP_012927176.1 | 1 - 2 | 363 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
