Assay Details
Gene Symbol: LOC101719334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843641.2 | XP_004843698.1 | 2 - 3 | 619 | 70 | |
XM_013071722.2 | XP_012927176.1 | 2 - 3 | 644 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
