Assay Details
| Gene Symbol: Faim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863008.3 | XP_004863065.1 | 9 - 10 | 962 | 59 | |
| XM_004863009.3 | XP_004863066.1 | 10 - 11 | 949 | 59 | ||
More Information
Set Membership: |
|

Faim2
Genomic Map