Assay Details
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866451.3 | XP_004866508.1 | 4 - 5 | 419 | 82 | |
XM_021245514.1 | XP_021101173.1 | 4 - 5 | 420 | 82 | ||
XM_021245515.1 | XP_021101174.1 | 3 - 4 | 216 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
