Assay Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865223.3 | XP_004865280.1 | 5 - 6 | 986 | 72 | |
| XM_021244659.1 | XP_021100318.1 | 5 - 6 | 1025 | 72 | ||
| XM_021244660.1 | XP_021100319.1 | 5 - 6 | 996 | 72 | ||
| XM_021244661.1 | XP_021100320.1 | 5 - 6 | 961 | 72 | ||
More Information
Set Membership: |
|

Blzf1
Genomic Map