Assay Details
Gene Symbol: LOC101707009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852778.2 | XP_004852835.1 | 2 - 3 | 299 | 110 | |
XM_021264822.1 | XP_021120481.1 | 2 - 3 | 300 | 110 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
