Assay Details
Gene Symbol: LOC101707009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852778.2 | XP_004852835.1 | 3 - 4 | 408 | 119 | |
XM_021264822.1 | XP_021120481.1 | 3 - 4 | 409 | 119 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
