Assay Details
Gene Symbol: Tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872867.3 | XP_004872924.1 | 2 - 3 | 1083 | 96 | |
XM_004872868.3 | XP_004872925.1 | 2 - 3 | 1090 | 96 | ||
XM_004872869.3 | XP_004872926.1 | 2 - 3 | 820 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
