Assay Details
| Gene Symbol: Ccbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239199.1 | XP_021094858.1 | 4 - 5 | 469 | 67 | |
| XM_021239200.1 | XP_021094859.1 | 4 - 5 | 469 | 67 | ||
More Information
Set Membership: |
|

Ccbe1
Genomic Map