Assay Details
Gene Symbol: Pop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850152.3 | XP_004850209.1 | 10 - 11 | 1621 | 62 | |
XM_021262677.1 | XP_021118336.1 | 10 - 11 | 1628 | 62 | ||
XM_021262678.1 | XP_021118337.1 | 9 - 10 | 1469 | 62 | ||
XM_021262679.1 | XP_021118338.1 | 8 - 9 | 1290 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
