Assay Details
Gene Symbol: LOC101712273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852572.2 | XP_004852629.1 | 1 - 2 | 1070 | 65 | |
XM_004852573.2 | XP_004852630.1 | 4 - 5 | 566 | 65 | ||
XM_004852574.2 | XP_004852631.1 | 4 - 5 | 505 | 65 | ||
XM_021264705.1 | XP_021120364.1 | 3 - 4 | 576 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
