Assay Details
Gene Symbol: St3gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837913.3 | XP_004837970.1 | 8 - 9 | 1716 | 61 | |
XM_004837914.3 | XP_004837971.1 | 9 - 10 | 1907 | 61 | ||
XM_004837915.3 | XP_004837972.1 | 7 - 8 | 1692 | 61 | ||
XM_004837916.3 | XP_004837973.1 | 8 - 9 | 1883 | 61 | ||
XM_004837917.3 | XP_004837974.1 | 8 - 9 | 1618 | 61 | ||
XM_004837918.3 | XP_004837975.1 | 8 - 9 | 1537 | 61 | ||
XM_013069595.2 | XP_012925049.1 | 9 - 10 | 1836 | 61 | ||
XM_013069596.2 | XP_012925050.1 | 10 - 11 | 2027 | 61 | ||
XM_013069597.2 | XP_012925051.1 | 7 - 8 | 1661 | 61 | ||
XM_021251715.1 | XP_021107374.1 | 6 - 7 | 1480 | 61 | ||
XM_021251736.1 | XP_021107395.1 | 8 - 9 | 1798 | 61 | ||
XM_021251739.1 | XP_021107398.1 | 8 - 9 | 1538 | 61 | ||
XM_021251742.1 | XP_021107401.1 | 10 - 11 | 2031 | 61 | ||
XM_021251761.1 | XP_021107420.1 | 8 - 9 | 1812 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
