Assay Details
Gene Symbol: Mre11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838047.2 | XP_004838104.1 | 16 - 17 | 2117 | 70 | |
XM_013069640.2 | XP_012925094.1 | 16 - 17 | 2117 | 70 | ||
XM_021252663.1 | XP_021108322.1 | 16 - 17 | 2068 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
