Assay Details
Gene Symbol: Pttg1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842510.3 | XP_004842567.1 | 2 - 3 | 239 | 60 | |
XM_021256396.1 | XP_021112055.1 | 2 - 3 | 669 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
