Assay Details
| Gene Symbol: Ndufb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834636.2 | XP_004834693.1 | 1 - 2 | 201 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb5| Gene Symbol: Ndufb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834636.2 | XP_004834693.1 | 1 - 2 | 201 | 64 | |
Set Membership: |
|