Assay Details
| Gene Symbol: Bri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840480.2 | XP_004840537.1 | 1 - 2 | 192 | 74 | |
| XM_004840481.2 | XP_004840538.1 | 1 - 2 | 178 | 74 | ||
| XM_021254768.1 | XP_021110427.1 | 1 - 2 | 193 | 74 | ||
| XM_021254771.1 | XP_021110430.1 | 1 - 2 | 190 | 74 | ||
More Information
Set Membership: |
|

Bri3
Genomic Map