Assay Details
| Gene Symbol: LOC101722119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856057.3 | XP_004856114.1 | 10 - 11 | 1184 | 96 | |
| XM_004856058.3 | XP_004856115.1 | 10 - 11 | 1191 | 96 | ||
| XM_013076445.2 | XP_012931899.1 | 10 - 11 | 1178 | 96 | ||
More Information
Set Membership: |
|

LOC101722119
Genomic Map