Assay Details
| Gene Symbol: Pkig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858186.3 | XP_004858243.1 | 3 - 4 | 445 | 75 | |
| XM_013077169.2 | XP_012932623.1 | 3 - 4 | 371 | 75 | ||
| XM_021238915.1 | XP_021094574.1 | 3 - 4 | 484 | 75 | ||
More Information
Set Membership: |
|

Pkig
Genomic Map