Assay Details
| Gene Symbol: Glrx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838719.2 | XP_004838776.1 | 10 - 11 | 1038 | 75 | |
| XM_021253257.1 | XP_021108916.1 | 10 - 11 | 958 | 75 | ||
More Information
Set Membership: |
|

Glrx3
Genomic Map