Assay Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835075.3 | XP_004835132.1 | 5 - 6 | 696 | 64 | |
| XM_004835076.3 | XP_004835133.1 | 5 - 6 | 696 | 64 | ||
| XM_004835077.3 | XP_004835134.1 | 5 - 6 | 696 | 64 | ||
| XM_021257686.1 | XP_021113345.1 | 5 - 6 | 696 | 64 | ||
| XM_021257693.1 | XP_021113352.1 | 5 - 6 | 907 | 64 | ||
| XM_021257699.1 | XP_021113358.1 | 5 - 6 | 879 | 64 | ||
| XM_021257713.1 | XP_021113372.1 | 5 - 6 | 696 | 64 | ||
| XM_021257723.1 | XP_021113382.1 | 5 - 6 | 696 | 64 | ||
| XM_021257730.1 | XP_021113389.1 | 5 - 6 | 696 | 64 | ||
| XM_021257737.1 | XP_021113396.1 | 5 - 6 | 696 | 64 | ||
| XM_021257747.1 | XP_021113406.1 | 5 - 6 | 696 | 64 | ||
More Information
Set Membership: |
|

Ano10
Genomic Map